Learn More
ATCC Streptococcus pneumoniae
Supplier:  ATCC 6303
Encompass
Streptococcus pneumoniae strain [CIP 104225] is a whole-genome sequenced bacterium that has applications in infectious disease research, respiratory disease research, and vaccine development. This strain is typed as serotype 3.
Catalog No. 50-238-4504