Learn More
ATCC Streptococcus pneumoniae; Strain S. Africa 6B-8
Supplier:  ATCC 700675
Encompass
This strain is resistant to penicillin. *Representative strain of the South African 6B-8 antibiotic-resistant pneumococcal clone.
Catalog No. 50-238-0584