Learn More
ATCC Streptococcus pneumoniae; Quantitative DNA
Supplier:  ATCC 700669DQ
Encompass
Quantitative Genomic DNA from Streptococcus pneumoniae strain Spain 23F-1 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load and for other molecular biology applications.
Catalog No. 50-238-3470