Learn More
ATCC Streptococcus pneumoniae ;hungary 19a-6
Supplier:  ATCC 700673
Encompass
Resistant to erythromycin, penicillin, tetracycline, chloramphenicol, and trimethoprim-sulfamethoxazole. Representative strain of the Pneumococcal Molecular Epidemiology Network (PMEN).
Catalog No. 50-238-0258