Learn More
ATCC Streptococcus pneumoniae; s. africa 19a-7
Catalog No. 502381543
Encompass
Intermediate resistance to penicillin. Representative strain of the Pneumococcal Molecular Epidemiology Network (PMEN).
Catalog No. 50-238-1543 Supplier ATCC Supplier No. 700674
Intermediate resistance to penicillin. Representative strain of the Pneumococcal Molecular Epidemiology Network (PMEN).