Learn More
Creative Biomart Recombinant HCV 4th Generation Antigen
Supplier:  Creative Biomart VAC0301
Encompass_Preferred
The E. coli derived HCV fourth generation antigen recombinant protein contains medium size core (55aa), NS3 (226aa), NS4 (3 epitopes) & NS5 (3 epitopes). HCV 4th Generation migrates at 46 kDa.
Catalog No. 50-208-5141