Learn More
ATCC Streptococcus pyogenes
Catalog No. 502382066
Encompass
Streptococcus pyogenes strain QC A62 is a whole-genome sequenced bacterium that was isolated from a human throat culture. This culture is typed as Lancefield’s group A and it can be used in emerging infectious disease research and as a quality control strain for BBL products.
Catalog No. 50-238-2066 Supplier ATCC Supplier No. 49399
Streptococcus pyogenes strain QC A62 is a whole-genome sequenced bacterium that was isolated from a human throat culture. This culture is typed as Lancefield’s group A and it can be used in emerging infectious disease research and as a quality control strain for BBL products.