Learn More
ATCC Leuconostoc citreum
Catalog No. 502379538
Encompass
Leuconostoc citreum strain 21 is a bacterial strain that is propagated in a 5% CO2 atmosphere in lactobacilli MRS medium.
Catalog No. 50-237-9538 Supplier ATCC Supplier No. 21437
Leuconostoc citreum strain 21 is a bacterial strain that is propagated in a 5% CO2 atmosphere in lactobacilli MRS medium.