Learn More
ACROBiosystems Streptococcus pyogenes IdeS Protein, His Tag
Catalog No. 502054836
Encompass_Preferred
Recombinant Protein;50UG;Streptococcus pyogenes IdeS, His Tag (IDS-S5143) is expressed from E.coli cells. It contains AA Asp 30 - Asn 339 (Accession # Q9F1R7-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.8 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).
This item is not returnable. View return policy
Catalog No. 50-205-4836 Supplier ACROBiosystems Supplier No. IDSS51432500U
Recombinant Protein;50UG;Streptococcus pyogenes IdeS, His Tag (IDS-S5143) is expressed from E.coli cells. It contains AA Asp 30 - Asn 339 (Accession # Q9F1R7-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.8 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE).