Learn More
ATCC Streptococcus pyogenes; SF370; genomic DNA
Catalog No. 502383625
Encompass
Genomic DNA isolated from Streptococcus pyogenes strain SF730; M1 GAS. This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3625 Supplier ATCC Supplier No. 700294D5
Genomic DNA isolated from Streptococcus pyogenes strain SF730; M1 GAS. This whole-genome sequenced product can be used in PCR and other molecular biology applications.