Learn More
ATCC Streptococcus pyogenes; Strain Bruno
Supplier:  ATCC 19615
Encompass
Streptococcus pyogenes strain Bruno [CIP 104226] is a whole-genome sequenced bacterial strain that was isolated from the pharynx of a child following an episode of a sore throat. This product has applications in media testing and it can be used as a control strain for Streptococcus Group A.
Catalog No. 50-238-2918
 Restricted Item