Learn More
ATCC Streptococcus pyogenes T1 genomic DNA
Catalog No. 502383928
Encompass
Genomic DNA isolated from Streptococcus pyogenes typing strain T1 (ATCC 12344). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3928 Supplier ATCC Supplier No. 12344D5
Genomic DNA isolated from Streptococcus pyogenes typing strain T1 (ATCC 12344). This whole-genome sequenced product can be used in PCR and other molecular biology applications.