Learn More
ATCC Streptococcus salivarius; subsp. salivarius
Catalog No. 502383803
Encompass
Streptococcus salivarius subsp. salivarius strain K-12 [DSM 13084] is a whole-genome sequenced clinical specimen that was isolated in 1989 from a healthy child.
Catalog No. 50-238-3803 Supplier ATCC Supplier No. BAA1024
Add to Cart
Add to Cart
Streptococcus salivarius subsp. salivarius strain K-12 [DSM 13084] is a whole-genome sequenced clinical specimen that was isolated in 1989 from a healthy child.