Learn More
ATCC Moraxella catarrhalis; Strain 61240
Catalog No. 502380061
Encompass
Moraxella catarrhalis strain 61240 was isolated in 2005 in Canada. This strain is part of the national surveillance study called CROSS (Canadian Respiratory Organism Susceptibility Study).
Catalog No. 50-238-0061 Supplier ATCC Supplier No. BAA1425
Moraxella catarrhalis strain 61240 was isolated in 2005 in Canada. This strain is part of the national surveillance study called CROSS (Canadian Respiratory Organism Susceptibility Study).