Learn More
ATCC Borrelia garinii; subsp. bavariensis; Strain PBi
Catalog No. 502380517
Encompass
Borrelia garinii subsp. bavariensis strain PBi is a whole-genome sequenced bacterial type strain that was isolated from cerebrospinal fluid. This strain does not grow on agar.
Catalog No. 50-238-0517 Supplier ATCC Supplier No. BAA2496
Borrelia garinii subsp. bavariensis strain PBi is a whole-genome sequenced bacterial type strain that was isolated from cerebrospinal fluid. This strain does not grow on agar.