Learn More
ATCC Methylorubrum extorquens; Strain NBRC 15911
Catalog No. 502380171
Encompass
Methylorubrum extorquens strain NBRC 15911 is a whole-genome sequenced bacterium that can be used for media growth promotion testing of R2A agar medium.
Catalog No. 50-238-0171 Supplier ATCC Supplier No. BAA2500
Methylorubrum extorquens strain NBRC 15911 is a whole-genome sequenced bacterium that can be used for media growth promotion testing of R2A agar medium.