Learn More
ATCC Streptococcus thermophilus
Supplier:  ATCC 19258
Encompass
Streptococcus thermophilus strain NCDO 573 is a whole-genome sequenced bacterial type strain that was isolated from pasteurized milk. This strain produces aminopeptidase N and has applications in quality control.
Catalog No. 50-238-2426
 Restricted Item