Learn More
Cayman Chemical StreptogramIn B 10mg
SDP
Catalog No. 502939872
Encompass_Preferred
A macrolide antibiotic; inhibits the growth of Clostridium, Peptostreptococcus, and Propionibacterium bacterial species (MIC90s = 2, 0.5, and 8 μg/ml, respectively); inhibits protein synthesis by interacting with the 50S ribosome
Catalog No. 50-293-9872 Supplier Cayman Chemical Supplier No. 2808610 MG
A macrolide antibiotic; inhibits the growth of Clostridium, Peptostreptococcus, and Propionibacterium bacterial species (MIC90s = 2, 0.5, and 8 μg/ml, respectively); inhibits protein synthesis by interacting with the 50S ribosome