Learn More
Alkali Scientific Streptomyces Agar w/3 wt% Sea Salt 30mL Plate, 10/Pk
SDP
Catalog No. 5000662970
Encompass_Preferred
Streptomyces Agar with sea salt is a specialized culture medium designed for the isolation and cultivation of marine actinomycetes. The formulation includes approximately 3% sea salt to mimic natural marine conditions, providing the ionic balance required for salt tolerant microorganisms. This environment supports the growth of marine Streptomyces species while inhibiting many non adapted terrestrial organisms. The 30mL plate format provides sufficient depth to support extended incubation periods often required for actinomycete growth. This medium is widely used in environmental microbiology and research for isolating organisms that produce bioactive compounds such as antibiotics.
Catalog No. 50-006-62970 Supplier Alkali Scientific Supplier No. SSTP01
Streptomyces Agar with sea salt is a specialized culture medium designed for the isolation and cultivation of marine actinomycetes. The formulation includes approximately 3% sea salt to mimic natural marine conditions, providing the ionic balance required for salt tolerant microorganisms. This environment supports the growth of marine Streptomyces species while inhibiting many non adapted terrestrial organisms. The 30mL plate format provides sufficient depth to support extended incubation periods often required for actinomycete growth. This medium is widely used in environmental microbiology and research for isolating organisms that produce bioactive compounds such as antibiotics.