Learn More
ATCC Streptomyces venezuelae
Catalog No. 502382039
Encompass
Streptomyces venezuelae strain 3627 was isolated from soil in Florence, Italy. This strain produces 10-deoxymethynolide and methymycin and has applications in industrial biotechnology.
Catalog No. 50-238-2039 Supplier ATCC Supplier No. 15439
Streptomyces venezuelae strain 3627 was isolated from soil in Florence, Italy. This strain produces 10-deoxymethynolide and methymycin and has applications in industrial biotechnology.