Learn More
Apexbio Technology LLC STREPTOMYCIN-100MG
SDP
Catalog No. 5000568063
Encompass_Preferred
Streptomycin (CAS No 57-92-1) is an aminoglycoside antibiotic derived from Streptomyces griseus primarily used in biomedical research to study bacterial protein synthesis inhibition by binding to the 30S ribosomal subunit This interaction disrupts mRNA translation leading to cellular toxicity in susceptible bacteria It exhibits in vitro activity typically in the nanomolar to low micromolar range making it effective for targeting Gram-negative bacterial strains Its applications include use in cell models and animal studies to investigate antibacterial effects and in drug screening for identifying synergies with other antibiotics Streptomycin is crucial in research exploring mechanisms of antibiotic resistance and optimizing treatment regimens Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68063 Supplier Apexbio Technology LLC Supplier No. A9080100
Streptomycin (CAS No 57-92-1) is an aminoglycoside antibiotic derived from Streptomyces griseus primarily used in biomedical research to study bacterial protein synthesis inhibition by binding to the 30S ribosomal subunit This interaction disrupts mRNA translation leading to cellular toxicity in susceptible bacteria It exhibits in vitro activity typically in the nanomolar to low micromolar range making it effective for targeting Gram-negative bacterial strains Its applications include use in cell models and animal studies to investigate antibacterial effects and in drug screening for identifying synergies with other antibiotics Streptomycin is crucial in research exploring mechanisms of antibiotic resistance and optimizing treatment regimens Experimental concentrations typically depend on the specific research design