Learn More
Medchemexpress LLC Streptomycin | 57-92-1 | 99.9% | 1 G
SDP
Catalog No. 5000411337
Encompass_Preferred
Streptomycin is an effective antibiotic used in the research of tuberculosis (TB) against M. tuberculosis. It is also a bacteriocidal agent effective against a variety of bacterial infections. It strongly binds to nucleic acids, interfering with and blocking protein synthesis while still allowing RNA and DNA synthesis. Additionally, it functions as a common antibiotic in culture media and blocks stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against M. tuberculosis
  • Bacteriocidal agent for research on various bacterial infections
  • Interferes with and blocks protein synthesis
  • Allows RNA and DNA synthesis to continue
  • Common antibiotic in culture media
  • Acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes
Catalog No. 50-004-11337 Supplier Medchemexpress LLC Supplier No. HYB19061G
Streptomycin is an effective antibiotic used in the research of tuberculosis (TB) against M. tuberculosis. It is also a bacteriocidal agent effective against a variety of bacterial infections. It strongly binds to nucleic acids, interfering with and blocking protein synthesis while still allowing RNA and DNA synthesis. Additionally, it functions as a common antibiotic in culture media and blocks stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against M. tuberculosis
  • Bacteriocidal agent for research on various bacterial infections
  • Interferes with and blocks protein synthesis
  • Allows RNA and DNA synthesis to continue
  • Common antibiotic in culture media
  • Acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes