Learn More
Medchemexpress LLC Streptomycin | 57-92-1 | 99.9% | 500 MG
SDP
Catalog No. 5000411338
Encompass_Preferred
Streptomycin is an effective antibiotic against M. tuberculosis and is used for the research of tuberculosis (TB). It is also a bacteriocidal agent used for the research of various bacterial infections. Streptomycin binds strongly to nucleic acids, interfering with and blocking protein synthesis while allowing continued RNA and DNA synthesis. Additionally, it functions as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes, and is a common antibiotic in culture media.
  • Effective antibiotic against M. tuberculosis
  • Bacteriocidal agent for bacterial infections research
  • Interferes with and blocks protein synthesis
  • Allows continued RNA and DNA synthesis
  • Blocker of stretch-activated and mechanosensitive ion channels
Catalog No. 50-004-11338 Supplier Medchemexpress LLC Supplier No. HYB1906500MG
Streptomycin is an effective antibiotic against M. tuberculosis and is used for the research of tuberculosis (TB). It is also a bacteriocidal agent used for the research of various bacterial infections. Streptomycin binds strongly to nucleic acids, interfering with and blocking protein synthesis while allowing continued RNA and DNA synthesis. Additionally, it functions as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes, and is a common antibiotic in culture media.
  • Effective antibiotic against M. tuberculosis
  • Bacteriocidal agent for bacterial infections research
  • Interferes with and blocks protein synthesis
  • Allows continued RNA and DNA synthesis
  • Blocker of stretch-activated and mechanosensitive ion channels