Learn More
Medchemexpress LLC Streptomycin | 57-92-1 | MFCD00072108 | 99.9% | 1 ML
SDP
Catalog No. 5000411336
Encompass_Preferred
Streptomycin (Agrept) is an effective antibiotic used in tuberculosis (TB) research and against various bacterial infections. It functions by binding strongly to nucleic acids, thereby interfering with and blocking protein synthesis. Additionally, it can block stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against *M. tuberculosis*
  • Used for tuberculosis research
  • Bactericidal agent for various bacterial infections
  • Interferes with and blocks protein synthesis
  • Permits continued RNA and DNA synthesis
  • Blocks stretch-activated and mechanosensitive ion channels
Catalog No. 50-004-11336 Supplier Medchemexpress LLC Supplier No. HYB190610MM/1ML
Streptomycin (Agrept) is an effective antibiotic used in tuberculosis (TB) research and against various bacterial infections. It functions by binding strongly to nucleic acids, thereby interfering with and blocking protein synthesis. Additionally, it can block stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against *M. tuberculosis*
  • Used for tuberculosis research
  • Bactericidal agent for various bacterial infections
  • Interferes with and blocks protein synthesis
  • Permits continued RNA and DNA synthesis
  • Blocks stretch-activated and mechanosensitive ion channels