Learn More
Medchemexpress LLC Streptomycin | 57-92-1 | MFCD00072108 | 99.9% | 5 G
SDP
Catalog No. 5000411102
Encompass_Preferred
Streptomycin (Agrept) is an effective antibiotic against *M. tuberculosis* and is used for the research of tuberculosis (TB). It is also a bacteriocidal agent for the research of various bacterial infections. Streptomycin binds strongly to nucleic acids, interfering with and blocking protein synthesis while permitting continued RNA and DNA synthesis. As a common antibiotic in culture media, it also acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against *M. tuberculosis* for TB research.
  • Bacteriocidal agent for various bacterial infections.
  • Binds strongly to nucleic acids, blocking protein synthesis while allowing RNA and DNA synthesis.
  • Acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
Catalog No. 50-004-11102 Supplier Medchemexpress LLC Supplier No. HYB19065G
Streptomycin (Agrept) is an effective antibiotic against *M. tuberculosis* and is used for the research of tuberculosis (TB). It is also a bacteriocidal agent for the research of various bacterial infections. Streptomycin binds strongly to nucleic acids, interfering with and blocking protein synthesis while permitting continued RNA and DNA synthesis. As a common antibiotic in culture media, it also acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.
  • Effective antibiotic against *M. tuberculosis* for TB research.
  • Bacteriocidal agent for various bacterial infections.
  • Binds strongly to nucleic acids, blocking protein synthesis while allowing RNA and DNA synthesis.
  • Acts as a blocker of stretch-activated and mechanosensitive ion channels in neurons and cardiac myocytes.