Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldon™ Streptomycin Disks
SCI_ED

Catalog No. S28511 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S28511 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S28511 Supplier Aldon™ Supplier No. XD1365
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Determines antibiotic susceptibility of microorganisms.

  • Each cartridge contains 50 disks (10mcg) of antibiotics with reclosable cap
  • Disks are 6.35mm (0.25 in.) in diameter
  • DOT Info: Non-regulated

Specifications

Grade 7 to 14
Format Disk
Product Type Streptomycin Disks
Diameter (English) Ring 0.25 in.
Quantity 50 Disks of 10 mcg
Color White
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.