Learn More
Perkin Elmer US LLC Streptomycin ELISA Kit
Supplier:  Perkin Elmer US LLC FOOD101401
Encompass_Preferred
Streptomycin ELISA Kit is a competitive enzyme immunoassay for the quantitative analysis of streptomycin in milk, soured milk, buttermilk, milk powder, sour cream, cheese, butter, yogurt, curd, whey, meat, liver, kidney, urine, serum, feed, and honey.
Catalog No. NC1833966