Learn More
Abcam Streptomycin ELISA Kit, 96TEST
Supplier:  Abcam AB28522696TEST
Encompass_Preferred
Streptomycin ELISA Kit is a Competitive ELISA kit for the measurement of Streptomycin in Tissue, Milk samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5436