Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptomycin Solution, ∼1 mg/mL in 1 mM EDTA, MilliporeSigma™ Supelco™

Catalog No. 111014093 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4093 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4093 Supplier MilliporeSigma Supelco Supplier No. 8588610ML
Add to Cart
Add to Cart

Chemical structure: aminoglycoside

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 3810-74-0
Physical Form Liquid
Linear Formula C21H39N7O12 · 1.5H2 SO4
MDL Number MFCD00037023
Type Analytical Standard
Quantity 10 mL
Formula Weight 728.69
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 1 mg/mL in 1 mM EDTA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.