Learn More
Teknova 50mg/mL Streptomycin Solution. 25mL, Sterile.
SDP
Supplier:  Teknova S6525
Encompass
Premade antibiotic solutions are ready-to-use as a supplement for your tissue culture, bacteria media or other specific needs. Antibiotics are often used in cell culture to prevent contamination, maintain aseptic conditions, or select for cells containing genetic modifications.
Catalog No. 50-238-8109