Learn More
Gold Biotechnology Inc Streptomycin Sulfate 100 g
SDP
Catalog No. 5000562126
Encompass_Preferred
Streptomycin is an aminoglycoside antibiotic primarily effective against gram-negative bacteria and Mycobacterium species It is commonly used in cell culture media to prevent bacterial contamination It has also been used to precipitate nucleic acids during protein purification techniques and to study mechanisms of Streptomycin resistance
Catalog No. 50-005-62126 Supplier Gold Biotechnology Inc Supplier No. S150100
Streptomycin is an aminoglycoside antibiotic primarily effective against gram-negative bacteria and Mycobacterium species It is commonly used in cell culture media to prevent bacterial contamination It has also been used to precipitate nucleic acids during protein purification techniques and to study mechanisms of Streptomycin resistance