Learn More
Crescent Chemical Co Inc STREPTOMYCIN SULFATE
SDP
Supplier:  Crescent Chemical Co Inc SE35500.02
Encompass_Preferred
Streptomycin sulfate research grade, Ph. Eur. Min. 720 U/mg. Aminoglycoside antibiotic from Streptomyces griseus. Blocks the initiation complex and causes misreading on ribosomes in protein synthesis. Inhibits function only of the 30S subunit. Size - 100G Sto
Catalog No. 50-248-163