Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Streptomycin sulfate

Catalog No. AC455340050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455340050 5 g
AC455340250 25 g
AC455341000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC455340050 Supplier Thermo Scientific Chemicals Supplier No. 455340050
  • $58.50
    $44.65 / Each of 1

    Save $13.85

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Streptomycin sulfate, CAS # 3810-74-0, is the sulfate salt form of streptomycin, and an aminoglycoside with antibacterial activity. It also shows a protein synthesis inhibitory activity.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Streptomycin sulfate is an aminoglycoside sulfate salt derived from Streptomyces griseus that shows antibacterial activity.

• It inhibits initiation and causes misreading of rRNA in protein synthesis. This compound binds to the S12 protein of the bacterial 30S ribosomal subunit, thereby inhibiting peptide elongation and protein synthesis.

Application

• Streptomycin sulfate can be used to study the mechanisms of streptomycin resistance

• This compound can be used in laboratory applications combined with penicillin to inhibit bacterial contamination in in vitro cell culture

• Streptomycin sulfate shows antibiotic activity against Mycobacterium tuberculosis

• This compound can be used as a protein synthesis inhibitor

• It acts against gram-negative and gram-positive bacteria • Streptomycin sulfate also shows a fungicide activity

• It can be used to remove nucleic acids from a biological extract

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3810-74-0
Molecular Formula C42H84N14O36S3
Molecular Weight (g/mol) 1457.376
InChI Key QTENRWWVYAAPBI-FFCQDDOVSA-N
PubChem CID 124080941
IUPAC Name 2-[(1R,2S,3S,4S,5R,6S)-3-(diaminomethylideneamino)-4-[(2R,3S,4S,5S)-3-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(hydroxymethyl)-3-(methylamino)oxan-2-yl]oxy-4-formyl-4-hydroxy-5-methyloxolan-2-yl]oxy-2,5,6-trihydroxycyclohexyl]guanidine;2-[(1R,2S,3S,4R,5R,6R)-3-(
SMILES CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.CC1C(C(C(O1)OC2C(C(C(C(C2O)O)N=C(N)N)O)N=C(N)N)OC3C(C(C(C(O3)CO)O)O)NC)(C=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Type Streptomycin Sulfate
Loss on Drying 7% max.
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 1457.4
Chemical Name or Material Streptomycin sulfate
Hazard Category Warning
Hazard Statement GHS H Statement Suspected of damaging the unborn child. Harmful if swallowed. May cause an allergic skin reaction.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention. Obtain special instructions before use. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.