Learn More
Gold Biotechnology Inc Streptomycin Sulfate 100 mg/mL Solution 10 mL
SDP
Catalog No. NC1247817
Encompass
Streptomycin is an aminoglycoside antibiotic primarily effective against gram-negative bacteria and Mycobacterium species. It is commonly used in cell culture media to prevent bacterial contamination. It has also been used to precipitate nucleic acids during protein purification techniques and to study mechanisms of Streptomycin resistance.
Catalog No. NC1247817 Supplier Gold Biotechnology Inc Supplier No. S150SL10
Streptomycin is an aminoglycoside antibiotic primarily effective against gram-negative bacteria and Mycobacterium species. It is commonly used in cell culture media to prevent bacterial contamination. It has also been used to precipitate nucleic acids during protein purification techniques and to study mechanisms of Streptomycin resistance.