Learn More
Apexbio Technology LLC Streptomycin sulfate 3810-74-0 10mM (in 1mL H2O)
SDP
Catalog No. 5000092853
Encompass_Preferred
Streptomycin sulfate is an aminoglycoside antibiotic targeting the bacterial 30S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby suppressing bacterial proliferation Streptomycin sulfate exerts its biological activity primarily through inhibition of mRNA translation by binding to the 30S subunit which impairs initiation and elongation processes during protein synthesis and disrupts the ribosome mRNA complex Based on these pharmacological properties streptomycin sulfate holds research potential in microbiological experimentation susceptibility assays bacterial resistance profiling microbial genetics studies and screening of transgenic cells expressing antibiotic-resistance markers
Catalog No. 50-000-92853 Supplier Apexbio Technology LLC Supplier No. B20345.1
Streptomycin sulfate is an aminoglycoside antibiotic targeting the bacterial 30S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby suppressing bacterial proliferation Streptomycin sulfate exerts its biological activity primarily through inhibition of mRNA translation by binding to the 30S subunit which impairs initiation and elongation processes during protein synthesis and disrupts the ribosome mRNA complex Based on these pharmacological properties streptomycin sulfate holds research potential in microbiological experimentation susceptibility assays bacterial resistance profiling microbial genetics studies and screening of transgenic cells expressing antibiotic-resistance markers