Learn More
Medchemexpress LLC Streptomycin (sulfate) | 3810-74-0 | 98.4% | 10 G
SDP
Catalog No. 5000405924
Encompass_Preferred
Streptomycin (sulfate) is an aminoglycoside antibiotic that inhibits protein synthesis. It is classified as a bacterial antibiotic and is used in infection research and plant cell culture. This product is intended for laboratory chemical use and substance manufacturing.
  • Aminoglycoside antibiotic
  • Inhibits protein synthesis
  • Used in infection research
  • Suitable for plant cell culture
  • For laboratory chemical applications
Catalog No. 50-004-05924 Supplier Medchemexpress LLC Supplier No. HYB047210G
Streptomycin (sulfate) is an aminoglycoside antibiotic that inhibits protein synthesis. It is classified as a bacterial antibiotic and is used in infection research and plant cell culture. This product is intended for laboratory chemical use and substance manufacturing.
  • Aminoglycoside antibiotic
  • Inhibits protein synthesis
  • Used in infection research
  • Suitable for plant cell culture
  • For laboratory chemical applications