Learn More
Medchemexpress LLC Streptomycin (sulfate) | 3810-74-0 | 98.4% | 50 G
SDP
Catalog No. 5000407741
Encompass_Preferred
Streptomycin sulfate is an aminoglycoside antibiotic that inhibits protein synthesis.
  • Aminoglycoside antibiotic
  • Inhibits protein synthesis
Catalog No. 50-004-07741 Supplier Medchemexpress LLC Supplier No. HYB047250G
Streptomycin sulfate is an aminoglycoside antibiotic that inhibits protein synthesis.
  • Aminoglycoside antibiotic
  • Inhibits protein synthesis