Learn More
Medchemexpress LLC Streptomycin sulfate | 3810-74-0 | MFCD00037023 | 98.4% | 1 ML
SDP
Catalog No. 5000405942
Encompass_Preferred
Streptomycin sulfate is an aminoglycoside antibiotic that inhibits protein synthesis. It is intended for research use only.
  • Inhibits protein synthesis.
  • Available as a 10 mM in 1 mL solution for immediate use.
  • High purity of 98.4%.
  • Referenced in numerous scientific publications.
  • Active against various bacterial strains.
Catalog No. 50-004-05942 Supplier Medchemexpress LLC Supplier No. HYB047210MM/1ML
Streptomycin sulfate is an aminoglycoside antibiotic that inhibits protein synthesis. It is intended for research use only.
  • Inhibits protein synthesis.
  • Available as a 10 mM in 1 mL solution for immediate use.
  • High purity of 98.4%.
  • Referenced in numerous scientific publications.
  • Active against various bacterial strains.