Learn More
Gold Biotechnology Inc Streptomycin Sulfate, USP Grade 50 g
SDP
Catalog No. 501532836
Encompass_Preferred
Streptomycin is an aminoglycoside antibiotic primarily effective against gram-negative bacteria and Mycobacterium species. It is commonly used in cell culture media to prevent bacterial contamination. It has also been used to precipitate nucleic acids during protein purification techniques and to study mechanisms of Streptomycin resistance.
Catalog No. 50-153-2836 Supplier Gold Biotechnology Inc Supplier No. S15050
Streptomycin is an aminoglycoside antibiotic primarily effective against gram-negative bacteria and Mycobacterium species. It is commonly used in cell culture media to prevent bacterial contamination. It has also been used to precipitate nucleic acids during protein purification techniques and to study mechanisms of Streptomycin resistance.