Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptomycin Sulphate, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139838
Change view
Click to view available options
Quantity:
500 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139838 500 g
S000036 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139838 Supplier G-Biosciences Supplier No. RC869
Add to Cart
Add to Cart

Streptomycin Sulfate is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.

It binds to the protein on the ribosomal 30S subunit and inhibits protein synthesis.

Chemical Identifiers

CAS 3810-74-0

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Sterility Non-sterile
Color White
Quantity 500 g
Percent Purity >95%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.