Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Streptonigrin | 3930-19-6 | 99.2% | 506.46 g/mol | C25H22N4O8 | 1MG
SDP

Catalog No. 5000237330
Encompass_Preferred

Streptonigrin is an aminoquinone antibiotic and protein-arginine deiminase (PAD) inhibitor supplied for research use. It is provided as a solid in mg-scale quantities, characterized by high purity and commonly used in biochemical and pharmacological studies investigating antimicrobial and enzymatic inhibition effects.

  • High purity of 99.2%.
  • Molecular weight 506.46 g/mol.
  • Chemical formula C25H22N4O8.
  • Supplied in small mg quantities suitable for laboratory research.
  • Recommended storage: powder at -20°C (long-term) and in solvent at -80°C (long-term).
  • Used as an antibiotic and PAD inhibitor in biochemical assays.

Catalog No. 50-002-37330 Supplier Medchemexpress LLC Supplier No. HY1245861MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Streptonigrin is an aminoquinone antibiotic and protein-arginine deiminase (PAD) inhibitor supplied for research use. It is provided as a solid in mg-scale quantities, characterized by high purity and commonly used in biochemical and pharmacological studies investigating antimicrobial and enzymatic inhibition effects.

  • High purity of 99.2%.
  • Molecular weight 506.46 g/mol.
  • Chemical formula C25H22N4O8.
  • Supplied in small mg quantities suitable for laboratory research.
  • Recommended storage: powder at -20°C (long-term) and in solvent at -80°C (long-term).
  • Used as an antibiotic and PAD inhibitor in biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.