Learn More
Enzo Life Sciences Streptothricin . sulfate (5mg). CAS: 96736-11-7
SDP
Supplier:  Enzo Life Sciences ALX380025M005
Encompass_Preferred
Antibiotic. Active against both Gram-positive and Gram-negative bacteria and some fungi. Alternative name: Nourseothricin sulfate. Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8914