Learn More
Apexbio Technology LLC Streptozocin, 18883-66-4, MFCD00006607, 100 mg
SDP
Catalog No. 501010150
Encompass_Preferred
MF: C8H15N3O7, MW: 265.22, Solubility: 10.3mg/mL in DMSO, 53.2mg/mL in H2O, 26.5mg/mL in EtOH with gentle warming. Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic beta-cells in vivo.
Catalog No. 50-101-0150 Supplier Apexbio Technology LLC Supplier No. A4457100
MF: C8H15N3O7, MW: 265.22, Solubility: 10.3mg/mL in DMSO, 53.2mg/mL in H2O, 26.5mg/mL in EtOH with gentle warming. Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic beta-cells in vivo.