Learn More
Medchemexpress LLC HY-13753 500mg , Streptozocin Streptozotocin;U 9889 CAS:18883-66-4 Purity:98%
SDP
Catalog No. 501492350
Encompass_Preferred
Medchemexpress, HY-13753 500mg Streptozocin Streptozotocin;U 9889 CAS:18883-66-4 Formula:C8H15N3O7 DNA alkylator Purity:98% Streptozocin is a potent DNA-methylating agent, with IC 50 values of 11.7, 904 and 1024 μg/mL for HL60, K562 and C1498 cells respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2350 Supplier Medchemexpress LLC Supplier No. HY13753500MG
Medchemexpress, HY-13753 500mg Streptozocin Streptozotocin;U 9889 CAS:18883-66-4 Formula:C8H15N3O7 DNA alkylator Purity:98% Streptozocin is a potent DNA-methylating agent, with IC 50 values of 11.7, 904 and 1024 μg/mL for HL60, K562 and C1498 cells respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.