Learn More
Abcam Streptozocin, Pancreatic beta-cell to x in, 100MG
Catalog No. 502706307
Encompass_Preferred
MW 265.22 Da, Purity >95%. Selective pancreatic β-cell toxin. DNA alkylating agent. Diabetagen and anticancer activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6307 Supplier Abcam Supplier No. AB142155100MG
MW 265.22 Da, Purity >95%. Selective pancreatic β-cell toxin. DNA alkylating agent. Diabetagen and anticancer activity in vivo.
The product is subject to the following: Abcam Restricted Use Statement