Learn More
Tocris Bioscience™ Streptozocin
DNA alkylator; antitumor and induces diabetes
Supplier: Tocris Bioscience™ 1621/500
Description
Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic β-cells in vivo.By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.