Learn More
Tocris Bioscience™ Streptozocin
GREENER_CHOICE
DNA alkylator; antitumor and induces diabetes
$314.22
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
16-215-00
GREENER_CHOICE
View Documents
Tocris Bioscience™
1621/500
Each of 1 for $314.22
Only null left
Add to Cart
Description
Description
Antibiotic and antitumor agent. Alkylates DNA and induces diabetes mellitus via reduction of nicotinamide adenine dinucleotide in pancreatic β-cells in vivo.