Learn More
Enzo Life Sciences Streptozotocin (100mg). CAS: 18883-66-4
SDP
Supplier:  Enzo Life Sciences ALX3800105100
Encompass_Preferred
Widely used diabetes inducer in rodents. Inhibition of β-cell O-GlcNAcase by streptozotocin is the mechanism that accounts for its diabetogenic toxicity. N-nitroso-containing antibiotic, acting as a nitric oxide (NO) donor and vasorelaxant. Relaxes phenylephrine-contracted aortic rings at 10µM. Potent methylating agent for DNA. Alternative name: Streptozocin, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose, STZ. Purity: ≥97% (Assay). Solubility: Soluble in water (nH2O at 102.8mg/ml) or 100% ethanol (200 proof at 0.92mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8904