Learn More
Cayman Chemical StreptozotocIn 100mg
SDP
Catalog No. 502953148
Encompass_Preferred
A glucosamine-nitrosourea which is commonly used to induce experimental diabetes in animals; specifically targets beta cells, entering via the glucose transporter GLUT2 and causing alkylation of DNA, leading to the destruction of beta cells
Catalog No. 50-295-3148 Supplier Cayman Chemical Supplier No. 13104100 MG
A glucosamine-nitrosourea which is commonly used to induce experimental diabetes in animals; specifically targets beta cells, entering via the glucose transporter GLUT2 and causing alkylation of DNA, leading to the destruction of beta cells