Learn More
Selleck Chemical LLC Streptozotocin (STZ) 50mg 18883-66-4 NSC-85998,Streptozocin
SDP
Catalog No. 501361133
Encompass_Preferred
Streptozotocin (STZ, NSC-85998, Streptozocin) is a glucosamine-nitrosourea derivative, which is a DNA-methylating, carcinogenic, antibiotic and diabetes inducing agent. Streptozotocin induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1133 Supplier Selleck Chemical LLC Supplier No. S131250MG
Streptozotocin (STZ, NSC-85998, Streptozocin) is a glucosamine-nitrosourea derivative, which is a DNA-methylating, carcinogenic, antibiotic and diabetes inducing agent. Streptozotocin induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.